L-tert-Leucinol 95%

Supplier: Thermo Scientific Chemicals
Warning

37753-0010 37753-0050
TS37753-0010EA 83 USD
TS37753-0010 TS37753-0050 TS37753-2500
L-tert-Leucinol 95%
L-tert-Leucinol
Formula: C₆H₁₅NO
Boiling Pt: 114…116 °C (10 mmHg)
Melting Pt: 30…34 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00192250
CAS Number: 112245-13-3

Order Now

Specification Test Results

Appearance Colorless to white Low melting mass or liquid
Infrared spectrum Conforms
GC ≥94 %
Specific optical rotation +34° to +40° (25°C, 589 nm) (c=1.5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR