Nocodazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl (6-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-yl)carbamate
35824-0100
35824-0500
TS35824-0100EA
106.25
USD
TS35824-0100
TS35824-0500
Nocodazole 98%
Nocodazole
Appearance: White to orange-brown Powder
Formula:
C₁₄H₁₁N₃O₃S MW: 301.32 g/mol Melting Pt: 300…305 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005588 CAS Number: 31430-18-9 |
Specification Test Results
Appearance | White to orange-brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...