MOPS-Na 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Sodium 4-morpholin-1-ylpropylsulfonate
, MOPS sodium salt
, Sodium 3-morpholinopropane-1-sulphonate
, 3-(N-Morpholino)propanesulphonic acid sodium salt
, 3-(N-Morpholino)propanesulfonic acid sodium salt
, Sodium 4-morpholin-1-ylpropylsulphonate
35259-0250
35259-0010
35259-1000
TS35259-1000EA
133
USD
TS35259-1000
TS35259-0010
TS35259-0250
MOPS-Na 98%
MOPS-Na
Formula:
C₇H₁₄NNaO₄S MW: 231.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064350 CAS Number: 71119-22-7 EINECS: 428-420-3 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Solubility | (23.125 g/l in cold water) Clear and colorless |
Solubility | complete dissolution within max 5 min |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...