X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: X-Gluc

33119-1000 33119-0250
TS33119-0250EA 166 USD
TS33119-0250 TS33119-1000
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%
X-GlcA ((5-Bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt)
Formula: C₂₀H₂₆BrClN₂O₇
MW: 521.79 g/mol
Melting Pt: 230 °C
Storage Temperature: Freezer
MDL Number: MFCD00058543
CAS Number: 114162-64-0

Order Now

Specification Test Results

Appearance Colorless to white to off-white Powder or crystals or needles
Infrared spectrum Conforms
Water ≤5 % (K.F.)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR