Calcimycin (A23187) free acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcium Ionophore A23187, Calcium Ionophore III, Antibiotic A23187

32808-0010 32808-0050
TS32808-0010EA 62 USD
TS32808-0010 TS32808-0050
Calcimycin (A23187) free acid 98%
Calcimycin (A23187)
Formula: C₂₉H₃₇N₃O₆
Melting Pt: 182…193 °C
Storage Temperature: Freezer
MDL Number: MFCD00151202
CAS Number: 52665-69-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 182 to 193 °C
TLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR