X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

32724-1000 32724-5000
TS32724-1000EA 98 USD
TS32724-1000 TS32724-5000
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99+% for molecular biology DNase-, RNase-, protease-free
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤0.2 %
TLC ≥99.0 %
Specific optical rotation -61° to -63° (20°C, 589 nm) (c=1, H2O/DMF 1/1)
Dnase,Rnase,Protease activity none detected

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR