6-Fluoroanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-6-fluorobenzoic acid

30645-0010 30645-0050
TS30645-0050EA 116 USD
TS30645-0050 TS30645-0010
6-Fluoroanthranilic acid 98%
6-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 170…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00067781
CAS Number: 434-76-4

Order Now

Specification Test Results

Appearance Yellow to beige to tan to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 165 to 171 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR