6-Chloroanthranilic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-6-chlorobenzoic acid

30050-0100 30050-0010
TS30050-0100EA 123 USD
TS30050-0100 TS30050-0010
6-Chloroanthranilic acid 99%
6-Chloroanthranilic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 143…147 °C
MDL Number: MFCD00051530
CAS Number: 2148-56-3
EINECS: 218-416-8

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 147 °C
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR