Phloroglucinol, anhydrous 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,5-Trihydroxybenzene

24176-0250 24176-1000 24176-5000
TS24176-0250EA 39 USD
TS24176-0250 TS24176-1000 TS24176-5000
Phloroglucinol, anhydrous 99+%
Phloroglucinol
Formula: C₆H₆O₃
Melting Pt: 215…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00002286
CAS Number: 108-73-6
EINECS: 203-611-2
Merck Index: 12,07482

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 213 to 218 °C
Loss on drying ≤2 % (1 g, 120 °C)
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR