4-(Aminomethyl)benzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

23222-0250 23222-1000
TS23222-0250EA 87 USD
TS23222-0250 TS23222-1000
4-(Aminomethyl)benzoic acid 99%
4-(Aminomethyl)benzoic acid
Formula: C₈H₉NO₂
Melting Pt: >300 °C
Flash Pt: >300 °C
MDL Number: MFCD00010203
CAS Number: 56-91-7
EINECS: 200-297-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤6 % (K.F.)
HPLC ≥98.5 %
Titration with HClO4 98.5 to 101.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR