L(-)-Carnitine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Levocarnitine hydrochloride, L-Carnitine hydrochloride, (R)-3-Carboxy-2-hydroxy-N,N,N-trimethylpropan-1-aminium chloride, LC-80, (R)-3-Hydroxy-4-(trimethylammonio)butyrate hydrochloride, L-Carnitine chloride, (R)-Carnitine hydrochloride, Levocarnitine chloride

23028-0050 23028-0250 23028-1000
TS23028-0050EA 48 USD
TS23028-0050 TS23028-0250 TS23028-1000
L(-)-Carnitine hydrochloride 98%
L(-)-Carnitine hydrochloride
Formula: C₇H₁₆ClNO₃
MW: 197.66 g/mol
Melting Pt: 142 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00066100
CAS Number: 6645-46-1
EINECS: 229-663-6

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
Sulfated ash ≤0.5 %
Specific optical rotation -21° to -24° (20°C, 589 nm) (c=1, H2O)
Titration with NaOH 97.5 to 102.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR