X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
BCIG
, X-GAL
, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
22777-5000
22777-1000
22777-0100
22777-0025
TS22777-0025EA
995
USD
TS22777-0025
TS22777-5000
TS22777-1000
TS22777-0100
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) 99%, pure
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula:
C₁₄H₁₅BrClNO₆ MW: 408.63 g/mol Melting Pt: 230 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005666 CAS Number: 7240-90-6 EINECS: 230-640-8 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤1 % (K.F.) |
HPLC | ≥98.5 % |
Specific optical rotation | -60° to -64° (20°C, 589 nm) (c=1, H2O/DMF 1/1) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...