Propylamine hydrochloride 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Aminopropane Hydrochloride
22189-0250
22189-1000
TS22189-0250EA
63.1
USD
TS22189-0250
TS22189-1000
Propylamine hydrochloride 99+%
Propylamine hydrochloride
Appearance: White to light beige Crystalline powder
Formula:
C₃H₉N MW: 95.57 g/mol Melting Pt: 160…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012911 CAS Number: 556-53-6 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 160 to 164 °C |
Titration with HClO4 | ≥99.0 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...