N-Phenyl-p-phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Warning

22100-0010 22100-1000 22100-2500
TS22100-0010EA 361 USD
TS22100-0010 TS22100-1000 TS22100-2500
N-Phenyl-p-phenylenediamine 98%
N-Phenyl-p-phenylenediamine
Formula: C₁₂H₁₂N₂
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 69…75 °C
Flash Pt: >250 °C
MDL Number: MFCD00007850
CAS Number: 101-54-2
EINECS: 202-951-9
UN: 3077

Order Now

Specification Test Results

Appearance Grey to brown to purple Powder or crystals or flakes or mass and/or chunks
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR