Ponceau Xylidine, pure

Supplier: Thermo Scientific Chemicals
Warning

19026-0250 19026-1000
TS19026-0250EA 80 USD
TS19026-0250 TS19026-1000
Ponceau Xylidine, pure
Ponceau Xylidine
Formula: C₁₈H₁₆N₂O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00003897
CAS Number: 3761-53-3

Order Now

Specification Test Results

Appearance Dark red to bordeaux Powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR