1-Ethylpropylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Pentylamine, 3-Aminopentane, Pentan-3-amine

18370-0050 18370-0250 18370-1000
TS18370-0050EA 63 USD
TS18370-0050 TS18370-0250 TS18370-1000
1-Ethylpropylamine 98+%
1-Ethylpropylamine
Formula: C₅H₁₃N
MW: 87.16 g/mol
Boiling Pt: 90
Density: 0.74 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008096
CAS Number: 616-24-0
UN: 1106
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR