5-Aminoisophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

18133-0250 18133-1000
TS18133-0250EA 49 USD
TS18133-0250 TS18133-1000
5-Aminoisophthalic acid 98%
5-Aminoisophthalic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 515
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002522
CAS Number: 99-31-0
EINECS: 202-748-5

Order Now

Specification Test Results

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
HPLC ≥94 %
Specific optical rotation -143° to -153° (20°C, 589 nm) (1% in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR