1-Adamantanemethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Adamantan-1-ylmethanamine, 1-Aminomethyladamantane
17742-0010
17742-0050
TS17742-0010EA
135
USD
TS17742-0010
TS17742-0050
1-Adamantanemethylamine 98%
1-Adamantanemethylamine
Appearance: Clear colorless to light yellow Liquid
Formula:
C₁₁H₁₉N MW: 165.28 g/mol Boiling Pt: 83…85 °C (.3 mmHg) Density: 0.93 g/cm³ Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074750 CAS Number: 17768-41-1 UN: 2735 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5127 to 1.5147 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...