γ-(N-Morpholino)propanesulfonic acid (MOPS) 99% for biochemistry
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Morpholinopropane-1-sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
17263-0025
17263-0250
17263-1000
17263-5000
TS17263-0025EA
1374
USD
TS17263-0025
TS17263-0250
TS17263-1000
TS17263-5000
γ-(N-Morpholino)propanesulfonic acid (MOPS) 99% for biochemistry
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula:
C₇H₁₅NO₄S MW: 209.27 g/mol Melting Pt: 277…282 °C Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006183 CAS Number: 1132-61-2 EINECS: 214-478-5 Merck Index: 13,06288 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1.0 % |
UV | at 280 nm A: ≤0.03 |
pH | 3.0 to 4.5 (1 % in water at 25°C) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...