Hydroxylammonium chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydroxylamine hydrochlorideHydroxylamine HCl

17036-5000 17036-0050 17036-0010 17036-1000
TS17036-0010EA 167 USD
TS17036-0010 TS17036-1000 TS17036-0050 TS17036-5000
Hydroxylammonium chloride 97%
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Melting Pt: 155…158 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 151 to 157 °C (Dec)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR