Diethyl pyrocarbonate 97%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
17025-0050
17025-0250
17025-1000
17025-5000
TS17025-0050EA
40
USD
TS17025-0050
TS17025-0250
TS17025-1000
TS17025-5000
Diethyl pyrocarbonate 97%, pure
Diethyl pyrocarbonate
Formula:
O(COOC₂H₅)₂ Boiling Pt: 93…94 °C (18 mmHg) Density: 1.1175 g/cm³ Flash Pt: 69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009106 CAS Number: 1609-47-8 EINECS: 216-542-8 Merck Index: 13,08088 |
Specification Test Results
Appearance | Clear colorless to amber Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3960 to 1.4000 (20°C, 589 nm) |
Specific gravity | (20/4°C) 1.11 to 1.25 |
Titration excess NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...