Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
16887-0025
16887-1000
16887-5000
TS16887-0025EA
2094
USD
TS16887-0025
TS16887-1000
TS16887-5000
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)
Formula:
C₈H₁₉NO₅ MW: 209.24 g/mol Melting Pt: 103…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002853 CAS Number: 6976-37-0 EINECS: 230-237-7 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤1.0 % (K.F.) |
Titration with HClO4 | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...