3-Nitro-p-toluic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-3-nitrobenzoic acid

16599-5000 16599-0250 16599-1000
TS16599-5000EA 327 USD
TS16599-5000 TS16599-0250 TS16599-1000
3-Nitro-p-toluic acid 99%
3-Nitro-p-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 187…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00007174
CAS Number: 96-98-0
EINECS: 202-549-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 190 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR