Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol

16382-0025 16382-1000 16382-5000
TS16382-0025EA 1389 USD
TS16382-0025 TS16382-1000 TS16382-5000
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 98%
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)
Formula: C₈H₁₉NO₅
MW: 209.24 g/mol
Melting Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002853
CAS Number: 6976-37-0
EINECS: 230-237-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR