(1S)-(-)-Borneol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol(-)-BorneolL(-)-Borneol

16065-0250 16065-5000 16065-1000
TS16065-0250EA 58 USD
TS16065-0250 TS16065-5000 TS16065-1000
(1S)-(-)-Borneol 97%
(1S)-(-)-Borneol
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 204…208 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00003759
CAS Number: 464-45-9
EINECS: 207-353-1
UN: 1312
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 204 to 208 °C
GC ≥96.0 %
Specific optical rotation -34.7° to -37.7° (20 °C, 589 nm) (c=5, C2H5OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR