3-Nitrophthalic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

16016-0025 16016-1000 16016-5000
TS16016-0025EA 854 USD
TS16016-0025 TS16016-1000 TS16016-5000
3-Nitrophthalic acid 95%
3-Nitrophthalic acid
Formula: C₈H₅NO₆
MW: 211.13 g/mol
Boiling Pt: 441
Melting Pt: 213…216 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007138
CAS Number: 603-11-2
EINECS: 210-030-8

Order Now

Specification Test Results

Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
HPLC ≥94 %
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR