Acetylcholine chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Acetoxyethyl)trimethylammonium chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, 2-Acetoxy-N,N,N-trimethylethanaminium chloride

15917-0050 15917-1000 15917-5000
TS15917-0050EA 34 USD
TS15917-0050 TS15917-1000 TS15917-5000
Acetylcholine chloride 99%
Acetylcholine chloride
Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
Melting Pt: 149…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011698
CAS Number: 60-31-1
EINECS: 200-468-8
Merck Index: 13,00089

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 151 °C
Titration Argentometric ≥98.5 % (on dried basis)
Loss on drying ≤1.0 % (105 °C, 3 hrs)
Sulfated ash ≤0.2 %
pH 5.2 to 9.2 (5 % in water)
Titration after Ion exchange ≥98.5 %
Chloride (Cl) ≤0.02 %
Arsenic (As) ≤0.0001 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.01 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR