Fumaronitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

15740-0050 15740-0250 15740-1000
TS15740-0050EA 73 USD
TS15740-0050 TS15740-0250 TS15740-1000
Fumaronitrile 98%
Fumaronitrile
Formula: C₄H₂N₂
Boiling Pt: 186 °C (760 mmHg)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00001928
CAS Number: 764-42-1
EINECS: 212-122-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥97.5 %
Specific optical rotation +10° to +20° (20 °C, 589 nm) (c=4 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR