1-Methylpiperazine 99%

Supplier: Thermo Scientific Chemicals
Danger

15691-0025 15691-0050 15691-1000 15691-5000
TS15691-0025EA 491 USD
TS15691-0025 TS15691-0050 TS15691-1000 TS15691-5000
1-Methylpiperazine 99%
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138
Melting Pt: -6
Density: 0.903 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4645 to 1.4665 (20°C, 589 nm)
Water ≤1 % (K.F.)
GC ≥98.5 %
Color scale ≤20 APHA
Stabilizer 0.025% BHT
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.05 ppm
Cobalt (Co) ≤0.05 ppm
Copper (Cu) ≤0.05 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.5 ppm
Manganese (Mn) ≤0.05 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.5 ppm
Acidity (CH3COOH) ≤0.001 %
Peroxides ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR