2-Piperidinoethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

15211-0050 15211-1000 15211-5000
TS15211-0050EA 27 USD
TS15211-0050 TS15211-1000 TS15211-5000
2-Piperidinoethanol 99%
2-Piperidinoethanol
Formula: C₇H₁₅NO
Melting Pt: 16.7 °C
Flash Pt: 77 °C
MDL Number: MFCD00006512
CAS Number: 3040-44-6
EINECS: 221-244-6

Order Now

Specification Test Results

Appearance Clear light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4794 to 1.4814 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR