2,2,6,6-Tetramethylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Danger

15195-0100 15195-0250 15195-1000
TS15195-0100EA 75 USD
TS15195-0100 TS15195-0250 TS15195-1000
2,2,6,6-Tetramethylpiperidine 98%
2,2,6,6-Tetramethylpiperidine
Formula: C₉H₁₉N
MW: 141.26 g/mol
Boiling Pt: 152
Density: 0.83 g/cm³
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00005985
CAS Number: 768-66-1
EINECS: 212-199-3
UN: 1992
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4425 to 1.4465 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR