Diphenylamine 99%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Phenylbenzeneamine
, N-Phenylaniline
15074-0010
15074-0051
15074-1000
15074-0050
TS15074-1000EA
41.76
USD
TS15074-1000
TS15074-0010
TS15074-0051
TS15074-0050
Diphenylamine 99%, pure
Diphenylamine
Formula:
(C₆H₅)₂NH MW: 169.23 g/mol Boiling Pt: 302 °C (760 mmHg) Melting Pt: 52…54 °C Flash Pt: 152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003014 CAS Number: 122-39-4 EINECS: 204-539-4 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to yellow to beige Flakes |
Infrared spectrum | Conforms |
Melting point | 52 to 55 °C |
GC | ≥98.5 % |
Free acid | ≤1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...