Hippuric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

15027-0025 15027-0050 15027-1000 15027-5000
TS15027-0025EA 406 USD
TS15027-0025 TS15027-0050 TS15027-1000 TS15027-5000
Hippuric acid 98%
Hippuric acid
Formula: C₆H₅C(O)NHCH₂COOH
Melting Pt: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00002692
CAS Number: 495-69-2
EINECS: 207-806-3
Merck Index: 13,04735

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 188 to 191 °C
Loss on drying ≤1 % (105 °C)
HPLC ≤1 % (Benzoic acid)
Titration with NaOH 97.5 to 102.5 % (On dried substance)
Chloride (Cl) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR