4-Aminobenzoic acid 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1
14621-0010
14621-0050
14621-2500
TS14621-0010EA
146
USD
TS14621-0010
TS14621-0050
TS14621-2500
4-Aminobenzoic acid 99%
4-Aminobenzoic acid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 340 Melting Pt: 188 Storage Temperature: Ambient |
MDL Number:
MFCD00007894 CAS Number: 150-13-0 EINECS: 205-753-0 Merck Index: 13,00422 |
Specification Test Results
Appearance | White to light yellow to light grey to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 186 to 189 °C |
Titration with NaOH | 98.5 to 101.5 % |
Elemental Analysis | 13.32 to 14.00 % N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...