Spermidine 99%

Supplier: Thermo Scientific Chemicals
Danger

13274-0010 13274-0050 13274-0250 13274-1000
TS13274-0010EA 55 USD
TS13274-0010 TS13274-0050 TS13274-0250 TS13274-1000
Spermidine 99%
Spermidine
Formula: C₇H₁₉N₃
Boiling Pt: 128…130 °C (14 mmHg)
Melting Pt: 22…25 °C
Density: 0.925 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
UN: 3259
ADR: 8,II
Merck Index: 12,08893

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4785 to 1.4805 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR