α-Hydroxy-o-tolyl-β-D-glucopyranoside 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol2-(Hydroxymethyl)phenyl-β-D-glucopyranosideSalicoside(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triolD(-)-SalicinD(−)-Salicin

13259-1000 13259-0050 13259-0250
TS13259-0050EA 50.74 USD
TS13259-0050 TS13259-1000 TS13259-0250
α-Hydroxy-o-tolyl-β-D-glucopyranoside 99+%
α-Hydroxy-o-tolyl β-D-glucopyranoside
Formula: C₁₃H₁₈O₇
MW: 286.28 g/mol
Melting Pt: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00006590
CAS Number: 138-52-3
EINECS: 205-331-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥99.0 %
Specific optical rotation -59.5° to -63.5° (20°C, 589 nm) (c=5, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR