4-Nitrobenzoic acid 99+%

Supplier: Thermo Scientific Chemicals
Warning

12846-1000 12846-0010 12846-0250
TS12846-0250EA 22.76 USD
TS12846-0250 TS12846-0010 TS12846-1000
4-Nitrobenzoic acid 99+%
4-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 239…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00007352
CAS Number: 62-23-7
EINECS: 200-526-2
Merck Index: 12,06686

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystalline powder or crystals
Infrared spectrum Conforms
Melting point 238 to 242 °C
HPLC ≥99.0 %
Titration with NaOH 99.0 to 101.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR