Indoline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydroindol

12224-0250 12224-1000 12224-5000
TS12224-0250EA 41 USD
TS12224-0250 TS12224-1000 TS12224-5000
Indoline 98%
Indoline
Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220
Melting Pt: -21
Density: 1.063 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005705
CAS Number: 496-15-1
EINECS: 207-816-8

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5950 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR