Guanidinium hydrochloride 98%, with anti-caking agent
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
12023-0010
12023-0025
12023-0100
12023-0251
12023-2500
TS12023-0010EA
100
USD
TS12023-0010
TS12023-0025
TS12023-0100
TS12023-0251
TS12023-2500
Guanidinium hydrochloride 98%, with anti-caking agent
Guanidinium hydrochloride
with <0.4% anticaking agent (SiO2)
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 179…186 °C Density: 1.34 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 179 to 186 °C |
Titration Argentometric | ≤0.5 % Ammonium chloride |
Water | ≤0.3 % |
Titration with HClO4 | ≥97.5 % |
Anticaking agent | ≤0.4 % (SiO2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...