Ethylenediamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane

11842-0025 11842-0100 11842-0010
TS11842-0025EA 133 USD
TS11842-0025 TS11842-0100 TS11842-0010
Ethylenediamine 99%
Ethylenediamine
Formula: C₂H₈N₂
MW: 60.1 g/mol
Boiling Pt: 117
Melting Pt: 8.5
Density: 0.898 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8 II
Merck Index: 13,03829

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4555 to 1.4575 (20°C, 589 nm)
Water ≤0.5 % (Coulometric)
GC ≥98.5 %
Sulfate (SO4) ≤0.02 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.002 %
Salicylic acid ≤0.04 % (HOC6H4COOH)
Insoluble matter ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR