Ethyl-4-(dimethylamino)benzoate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Dimethylamino)benzoic acid ethyl ester, Ethyl 4-dimethylaminobenzoate

11840-0050 11840-1000 11840-5000
TS11840-0050EA 25 USD
TS11840-0050 TS11840-1000 TS11840-5000
Ethyl-4-(dimethylamino)benzoate 99+%
Ethyl 4-(dimethylamino)benzoate
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 112
Melting Pt: 62…65 °C
Density: 1.06 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00009115
CAS Number: 10287-53-3
EINECS: 233-634-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 65 °C
GC ≥99.0 %
pH 5.0 to 7.0 (2 % solution at 25.0 °C)
Sensitivity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR