3-Iodo-5-nitrobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR401063-1G OR401063-250MG OR401063-5G
76621-294EA 33.34 USD
76621-294 76621-296 76621-298
3-Iodo-5-nitrobenzyl bromide 95%
3-Iodo-5-nitrobenzyl bromide
Formula: C₇H₅BrINO₂
Storage Temperature: Ambient
MDL Number: MFCD18398007
CAS Number: 1261644-60-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR