3-Bromo-5-methylbenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

OR400207-1G OR400207-250MG OR400207-5G
76621-170EA 192.01 USD
76621-170 76621-172 76621-174
3-Bromo-5-methylbenzyl bromide
3-Bromo-5-methylbenzyl bromide
Formula: C₈H₈Br₂
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD18204792
CAS Number: 51719-69-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR