5-(tert-Butyl)thiophene-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-(tert-Butyl)-2-thenoic acid, 5-(tert-Butyl)-2-carboxythiophene

OR01670-1G OR01670-5G
76610-836EA 43.8 USD
76610-836 76610-838 76610-840
5-(tert-Butyl)thiophene-2-carboxylic acid
5-(tert-Butyl)thiophene-2-carboxylic acid
Formula: C₉H₁₂O₂S
Melting Pt: 125…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01566583
CAS Number: 29212-25-7
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR