N-(1-Naphthyl)formamide 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR55038-25G OR55038-5G
76592-210EA 260.78 USD
76592-210 76592-208
N-(1-Naphthyl)formamide 99%
N-(1-Naphthyl)formamide
Formula: C₁₁H₉NO
Boiling Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00086455
CAS Number: 6330-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR