1-(4-Methylpyridin-2-yl)piperazine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR0889-1G OR0889-250MG OR0889-5G
76610-876EA 126.32 USD
76610-876 76610-878 76610-880
1-(4-Methylpyridin-2-yl)piperazine 97%
1-(4-Methylpyridin-2-yl)piperazine
Formula: C₁₀H₁₅N₃
Boiling Pt: 161…163 °C/8mmHg
Storage Temperature: Ambient
MDL Number: MFCD03412118
CAS Number: 34803-67-3
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR