1-Acetyl-2-methyl-5-indolinesulfonoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR3671-1G OR3671-5G
76611-748EA 154.84 USD
76611-748 76611-750
1-Acetyl-2-methyl-5-indolinesulfonoyl chloride
1-Acetyl-2-methyl-5-indolinesulfonoyl chloride
Formula: C₁₁H₁₂ClNO₃S
Melting Pt: 143…145 °C
Storage Temperature: Ambient
MDL Number: MFCD07841689
CAS Number: 841275-78-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR