2-Methyl-2H-pyrazole-3-sulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Methyl-1H-pyrazole-5-sulfonyl chloride1-Methyl-1H-pyrazole-5-sulphonyl chloride

OR17845-1G OR17845-250MG
76620-710EA 69.01 USD
76620-710 76620-708
2-Methyl-2H-pyrazole-3-sulfonyl chloride 95%
2-Methyl-2H-pyrazole-3-sulfonyl chloride
Formula: C₄H₅ClN₂O₂S
MW: 180.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09065021
CAS Number: 1020721-61-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR