Aminomalononitrile-p-toluenesulfonate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Aminomalononitrile p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulfonate, Aminomalononitrile p-Toluenesulphonic acid, Aminomalononitrile p-Toluenesulfonic acid, Dicyanomethylammonium p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulphonate, Aminomalononitrile p-Toluenesulfonate, Dicyanomethylammonium p-Toluenesulfonate

OR28794-25G OR28794-5G OR28794-10G
76625-040EA 101.88 USD
76625-040 76625-042 76625-038
Aminomalononitrile-p-toluenesulfonate 97%
Aminomalononitrile-p-toluenesulfonate
Formula: C₃H₃N₃
MW: 253.28 g/mol
Melting Pt: 169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012848
CAS Number: 5098-14-6
EINECS: 225-817-1
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR