Azetidine-3-carbonitrile hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: azetidine-3-carbonitrile hydrochloride

OR309045-1G OR309045-5G
76616-394EA 26.48 USD
76616-394 76616-396
Azetidine-3-carbonitrile hydrochloride 95%
Azetidine-3-carbonitrile hydrochloride
Formula: C₄H₇ClN₂
MW: 118.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749828
CAS Number: 345954-83-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR